2,6-Di-tert-butyl-p-kresol ≥99.0%, granular FCC
Leverantör: Spectrum Chemical
Synonyms: Butylated hydroxytoluene, Butylhydroxytoluene, 2,6-Di-tert-butyl-4-methylphenol, BHT
B1095-125GM B1095-2.5KG B1095-500GM
SPCMB1095-125GMEA 1675 SEK
SPCMB1095-125GM SPCMB1095-2.5KG SPCMB1095-500GM
2,6-Di-tert-butyl-p-kresol ≥99.0%, granular FCC
2,6-Di-tert-butyl-p-kresol
BHT, Granular, FCC is used in small quantities as a food additive as an antioxidant. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
GHS hazard : Exclamation mark.
Formel: C₁₅H₂₄O
MW: 220.35 g/mol
Kokpunkt: 265
Flampunkt: 127
Storage Temperature: Omgivande
MDL Number: MFCD00011644
CAS nummer: 128-37-0
Merck Index: 13,01547
Order Now
Specifikation testresultat
Assay (C₁₅H₂₄O) 99.0 % min.
Residue On Ignition max.0.002 %
Lead max. 2 mg/kg
Identification To Pass Test