Pyrroloquinoline quinone 95%

Leverantör: Thermo Scientific
Varning

Synonyms: PPQ

467010010 467012500
ACRO467010010EA 12350 SEK
ACRO467010010 ACRO467012500
Pyrroloquinoline quinone 95%
Pyrroloquinoline quinone
Formel: C₁₄H₆N₂O₈
MW: 330,21 g/mol
MDL Number: MFCD00043125
CAS nummer: 72909-34-3

Order Now

Specifikation testresultat

Appearance (Color) Dark orange to red to brown
Appearance (Form) powder
Infrared spectrum Conforms
HPLC ≥95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR