Battery Test Chambers with Safety Packages for Performance or Stress Tests
Leverantör: Binder
9418-0113 9419-0409 9420-0142 9421-0134 9421-0243 9421-0018 9421-0195 9421-0240 9421-0262
390-1678EA 0 SEK
390-1678 390-1679 390-1680 390-1682 390-1685 390-1681 390-1683 390-1684 390-1686
Battery Test Chambers with Safety Packages for Performance or Stress Tests
Testkammare Temperaturtestkammare
The test chambers for energy storage systems carrying out stress and performance tests, offer maximum user convenience and are exactly in line with EUCAR Hazard Level 4, 5 or 6. Within a manufacturing process, the KB series equipped with Package P, P Plus or Package S is perfectly suited for forming the cells.
  • Perfectly suited for temperature tests between 0 and 100 °C
  • Excellent price-performance ratio
  • Compact dimensions compared to other models
  • Small footprint – ideal for small setup areas
  • Choice of 3 different packages for performance and stress tests
Package P for performance tests, Hazard classification EUCAR hazard level 4/5/6 (Leakage, loss of mass > 50%, Fire or flame and Rupture respectively) includes:
•Class 2 independent temperature safety device when temperature is set to 120 °C
•Temperature range limited to 110 °C on the controller
•Stainless-steel pressure relief flap installed on top of the unit in the middle
•Enhanced door-locking mechanism with reinforced brackets
•Inert gas connections
Package P Plus for extended performance tests, Hazard classification EUCAR hazard level 5/6 (Fire or flame and Rupture respectively) includes:
•Class 2 independent temperature safety device when temperature is set to 120 °C
•Temperature range limited to 110 °C on the controller
•Stainless-steel pressure relief flap installed on top of the unit in the middle
•Enhanced door-locking mechanism with reinforced brackets
•Inert gas connections
•CO2 fire suppression equipment
Package S for stress tests (Hazard classification EUCAR hazard level 6 - Rupture) includes:
•Class 2 independent temperature safety device when temperature is set to 120 °C
•Temperature range limited to 110 °C on the controller
•Stainless-steel reversible pressure relief flap installed on top of the unit in the middle
•Enhanced door-locking mechanism with reinforced brackets
•Monitoring of CO, H2, and O2 in testing area atmosphere
•N2 permanent inertization
•CO2 flushing device
For details of the specification of Refrigerated incubators, KB series please see separate listing. Please note that Safety packages need to be ordered in addition with standard Binder KB cooled incubators and can not be retro-fitted to KB chambers that have already been purchased.