Tedizolid
Leverantör: Thermo Scientific
Synonyms: (R)-3-(3-Fluoro-4-(6-(2-methyl-2H-tetrazol-5-yl)pyridin-3-yl)phenyl)-5-(hydroxymethyl)oxazolidin-2-one
467580010 467582500
467580010.EA 4075 SEK
467580010. 467582500.
Tedizolid
Tedizolid
Formel: C₁₇H₁₅FN₆O₃
MW: 370,34 g/mol
Smältpunkt: 201°C
Storage Temperature: Kylskåp
MDL Number: MFCD19442562
CAS nummer: 856866-72-3
Order Now
Specifikation testresultat
Appearance White to beige powder
Infrared spectrum Conforms
HPLC ≥ 97.5%
Loss on drying ≤ 6%