Teriflunomide

Leverantör: Thermo Fisher Scientific
Varning

Synonyms: N-[4-(Trifluoromethyl)phenyl]-2-cyano-3-hydroxycrotonamide, (Z)-2-Cyano-3-hydroxy-N-[4-(trifluoromethyl)phenyl]-2-butenamide, (Z)-2-Cyano-3-hydroxy-N-(4-(trifluoromethyl)phenyl)but-2-enamide

467110010 467112500
ACRO467110010EA 5350 SEK
ACRO467110010 ACRO467112500
Teriflunomide
Teriflunomide
Formel: C₁₂H₉F₃N₂O₂
Smältpunkt: 229°C
Storage Temperature: Frys
MDL Number: MFCD00910058
CAS nummer: 163451-81-8

Order Now

Specifikation testresultat

Appearance White to almost white crystalline powder
Infrared spectrum Conforms
Melting point 227.0°C to 231.0°C
HPLC ≥ 99.0%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR