Rocuronium bromide 97%
Leverantör: Thermo Fisher Scientific
Varning
465020010 465022500
ACRO465020010EA 5075 SEK
ACRO465020010 ACRO465022500
Rocuronium bromide 97%
Rocuronium bromide
Formel: C₃₂H₅₃BrN₂O₄
MW: 609.69 g/mol
Smältpunkt: 162°C…164°C
Storage Temperature: Frys
MDL Number: MFCD00867768
CAS nummer: 119302-91-9
Order Now
Specifikation testresultat
Appearance White to light yellow to light brown crystalline powder
Infrared spectrum Conforms
Titration with HClO4 96.0 to 104.0% (dry substance)
Loss on drying ≤ 6%