Columbin ≥98%
Leverantör: Cayman Chemical
Synonyms:
9-(3-furanyl)-1R,4R,4aR,5,6,6aR,9S,10,10aS,10bS-decahydro-4-hydroxy-4a,10a-dimethyl-1,4-etheno-3H,7H-benzo[1,2-c:3,4-c']dipyran-3,7-dione
24105-1
24105-10
24105-25
24105-5
CAYM24105-1EA
1300
SEK
CAYM24105-1
CAYM24105-10
CAYM24105-25
CAYM24105-5
Columbin ≥98%
Columbin
Columbin is a diterpenoid furanolactone isolated from J. columba, A. albida, and T. bakis that has diverse biological activities.
Observera: 98
Formel:
C₂₀H₂₂O₆ MW: 358,4 g/mol Storage Temperature: Frys |
CAS nummer:
546-97-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...