Trikaliumfosfat 90+ %, ren
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Potassium phosphate tribasic, Potassium phosphate
467330010 467330050 467330250
ACRO467330010EA 855 SEK
ACRO467330010 ACRO467330050 ACRO467330250
Trikaliumfosfat 90+ %, ren
Trikaliumfosfat
Formel: K₃O₄P
MW: 212.27 g/mol
Smältpunkt: 1340 °C
Storage Temperature: Omgivande
MDL Number: MFCD00036295
CAS nummer: 7778-53-2
EINECS: 231-907-1
Merck Index: 12,07830
Order Now
Specifikation testresultat
Appearance (Color) White
Appearance (Form) Powder or granules and/or chunks
Titration excess HCl ≥90.0 %
Water =<10 %