Pentair Mixed-Bed Deionization Filter Cartridges
Leverantör: Avantor Fluid Handling
29830-63 29830-64 29830-65
MFLX29830-63EA 795 SEK
MFLX29830-63 MFLX29830-64 MFLX29830-65
Pentair Mixed-Bed Deionization Filter Cartridges
Filter Filterpatroner
Deionize and minimize the TOC (total organic carbon) in water.
  • FDA-compliant
Deionization Cartridges are designed for deionizing water at 16 MΩ, the mixed-bed cartridges also minimize the TOC (total organic carbon) level. Liquid flows through a bed. Not a double or single open-end cartridge. Compatible with any 9-3/4" housing without a center rod (i.e. 01508-25 or VPN 150068). Not compatible with 01508-40 or 01508-41 (VPN 156017-02 or 156018-02).
All materials are FDA-compliant.