+GF+ Piping Systems Metering Ball Valves
Leverantör: Avantor Fluid Handling
01360-40 01360-41 01360-45
MFLX01360-40EA 1975 SEK
MFLX01360-40 MFLX01360-41 MFLX01360-45
+GF+ Piping Systems Metering Ball Valves
Ventiler Doseringsventiler
Precise control over 180° range.
  • Choose between PVC or polypropylene (PP) construction
Ball valves give you precise control over the entire 180° range. True union design lets you remove wetted parts. Valves have Viton® or EPDM body seals and seats. Less than 5" overall length. Calibrated in 5° increments.