Fenolftalein 1% ethanol_70 Indicator_pH
Leverantör: VWR Chemicals
Fara
Synonyms: 3,3-Bis(4-hydroxyphenyl)isobenzofuran-1(3H)-one
31723.230EA 565 SEK
31723.230 31723.290
Fenolftalein 1% ethanol_70 Indicator_pH
Fenolftalein
  • Transition range pH 8.2-9.8
Formel: C₂₀H₁₄O₄
MW: 318.33 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00005913
CAS nummer: 77-09-8
EINECS: 201-004-7
UN: 1170
ADR: 3 III
Merck Index: 14,07243
Order Now
Specifikation testresultat
Application test passes test