Benzyl 3-O-allyl-2-O-benzyl-4,6-O-benzylidene-beta-D-glucopyranoside
Leverantör: Apollo Scientific
BICL5020-100MG BICL5020-250MG BICL5020-500MG
APOSBICL5020-100MGEA 2575 SEK
APOSBICL5020-100MG APOSBICL5020-250MG APOSBICL5020-500MG
Benzyl 3-O-allyl-2-O-benzyl-4,6-O-benzylidene-beta-D-glucopyranoside
Benzyl 3-O-allyl-2-O-benzyl-4,6-O-benzylidene-beta-D-glucopyranoside
Formel: C30H32O6
MW: 488.57 g/mol
Storage Temperature: Omgivande
CAS nummer: 103188-94-9