Nicotinamide adenine dinucleotide phosphate (reduced form) tetrasodium salt ≥98%
Leverantör: Apollo Scientific
Varning
BIB3014-100MG BIB3014-500MG
APOSBIB3014-100MGEA 720 SEK
APOSBIB3014-100MG APOSBIB3014-500MG
Nicotinamide adenine dinucleotide phosphate (reduced form) tetrasodium salt ≥98%
Nicotinamide adenine dinucleotide phosphate (reduced form) tetrasodium salt
Formel: C₂₁H₂₆N₇O₁₇P₃.4Na
MW: 833.348 g/mol
Storage Temperature: Frys
MDL Number: MFCD00036263
CAS nummer: 2646-71-1
EINECS: 220-163-3