β-Dihydronicotinamide adenine dinucleotide disodium salt (NADH-Na₂, reduced form) ≥98%
Leverantör: Apollo Scientific
Synonyms: Nicotinamide adenine dinucleotide reduced, disodium salt (reduced form), NADH disodium salt, ((2R,3S,4R,5R)-5-(6-Aminopurin-9-yl)-3,4-dihydroxy-oxolan-2-yl)methoxy-((((2R,3S,4R,5R)-5-(3-carbamoyl-4H-pyridin-1-yl)-3,4-dihydroxy-oxolan-2-yl)methoxy)hydroxyphosphoryl)oxyphosphinic acid disodium salt
BIB3012-1G BIB3012-5G BIB3012-25G
APOSBIB3012-1GEA 430 SEK
APOSBIB3012-1G APOSBIB3012-5G APOSBIB3012-25G
β-Dihydronicotinamide adenine dinucleotide disodium salt (NADH-Na₂, reduced form) ≥98%
β-Dihydronicotinamide adenine dinucleotide disodium salt (NADH-Na2, reduced form)
Formel: C₂₁H₂₇N₇Na₂O₁₄P₂
MW: 709.41 g/mol
Smältpunkt: 140…142 °C
Storage Temperature: Frys
MDL Number: MFCD00036200
CAS nummer: 606-68-8
EINECS: 210-123-3