2-Oxo-1-[3-(trifluoromethyl)benzyl]-1,2-dihydro-3-pyridine carboxylic acid 97%
Leverantör: Apollo Scientific
Varning
Synonyms: 1-[3-(Trifluoromethyl)benzyl]pyridin-2-one-3-carboxylic acid
PC9471-1G PC9471-100MG PC9471-250MG
APOSPC9471-1GEA 2150 SEK
APOSPC9471-1G APOSPC9471-100MG APOSPC9471-250MG
2-Oxo-1-[3-(trifluoromethyl)benzyl]-1,2-dihydro-3-pyridine carboxylic acid 97%
2-Oxo-1-[3-(trifluoromethyl)benzyl]-1,2-dihydro-3-pyridine carboxylic acid
Formel: C₁₄H₁₀F₃NO₃
Smältpunkt: 159…161 °C
Storage Temperature: Omgivande
MDL Number: MFCD00139983
CAS nummer: 338754-66-8