2-({2-Oxo-2-[3-(trifluoromethyl)anilino]ethyl}sulphanyl)acetic acid
Leverantör: Apollo Scientific
Synonyms: Thiodiacetic acid mono-3-(trifluoromethyl)anilide
PC9469-1G PC9469-500MG PC9469-5G
APOSPC9469-1GEA 1825 SEK
APOSPC9469-1G APOSPC9469-500MG APOSPC9469-5G
2-({2-Oxo-2-[3-(trifluoromethyl)anilino]ethyl}sulphanyl)acetic acid
2-({2-Oxo-2-[3-(trifluoromethyl)anilino]ethyl}sulphanyl)acetic acid
Formel: C₁₁H₁₀F₃NO₃S
Smältpunkt: 102…104 °C
Storage Temperature: Omgivande
MDL Number: MFCD00202305
CAS nummer: 338421-05-9