1-Methyl-3-(trifluoromethyl)-1H-pyrazole-4-carboxylic acid 98%
Leverantör: Apollo Scientific
Varning
Synonyms: 1-Methyl-3-(trifluoromethyl)pyrazole-4-carboxylic acid
PC9458-1G PC9458-5G
APOSPC9458-1GEA 210 SEK
APOSPC9458-1G APOSPC9458-5G
1-Methyl-3-(trifluoromethyl)-1H-pyrazole-4-carboxylic acid 98%
1-Methyl-3-(trifluoromethyl)-1H-pyrazole-4-carboxylic acid
Formel: C₆H₅F₃N₂O₂
MW: 194,1113 g/mol
Smältpunkt: 200…204 °C
Storage Temperature: Omgivande
MDL Number: MFCD01936005
CAS nummer: 113100-53-1