4-Hydroxy-6-(trifluoromethoxy)quinoline-3-carboxylic acid, Technical Grade
Leverantör: Apollo Scientific
Varning
PC9310-10G PC9310-50G
APOSPC9310-10GEA 2075 SEK
APOSPC9310-10G APOSPC9310-50G
4-Hydroxy-6-(trifluoromethoxy)quinoline-3-carboxylic acid, Technical Grade
4-Hydroxy-6-(trifluoromethoxy)quinoline-3-carboxylic acid
Formel: C₁₁H₆F₃NO₄
MW: 273.16 g/mol
Smältpunkt: 245 °C
Storage Temperature: Omgivande
MDL Number: MFCD00219848
CAS nummer: 175203-86-8