(2,4-Difluorophenoxy)acetic acid 98%
Leverantör: Apollo Scientific
PC911112-1G PC911112-5G
APOSPC911112-1GEA 2250 SEK
APOSPC911112-1G APOSPC911112-5G
(2,4-Difluorophenoxy)acetic acid 98%
(2,4-Difluorophenoxy)acetic acid
Formel: C₈H₆F₂O₃
Storage Temperature: Omgivande
MDL Number: MFCD03422203
CAS nummer: 399-44-0