2,4-Difluoro-5-nitroaniline 97%
Leverantör: Apollo Scientific
PC909005-1G
PC909005-250MG
APOSPC909005-1GEA
755
SEK
APOSPC909005-1G
APOSPC909005-250MG
2,4-Difluoro-5-nitroaniline 97%
2,4-Difluoro-5-nitroaniline
Formel:
C₆H₄F₂N₂O₂ MW: 174,11 g/mol Storage Temperature: Omgivande |
MDL Number:
MFCD04038395 CAS nummer: 123344-02-5 |