2-(3-Fluorophenoxy)ethylamine hydrochloride 98%
Leverantör: Apollo Scientific
Synonyms: 2-(3-Fluorophenoxy)ethylamine
PC908941-1G PC908941-5G
APOSPC908941-1GEA 2250 SEK
APOSPC908941-1G APOSPC908941-5G
2-(3-Fluorophenoxy)ethylamine hydrochloride 98%
2-(3-Fluorophenoxy)ethylamine hydrochloride
Formel: C₈H₁₁ClFNO
MW: 191,63 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD11099553
CAS nummer: 120351-93-1