2-Cyano-4'-fluoroacetanilide 95%

Leverantör: Apollo Scientific

Synonyms: 2-Cyano-N-(4-fluorophenyl)acetamide

PC908683-1G PC908683-5G
APOSPC908683-1GEA 925 SEK
APOSPC908683-1G APOSPC908683-5G
2-Cyano-4'-fluoroacetanilide 95%
2-Cyano-4'-fluoroacetanilide
Formel: C₉H₇FN₂O
MW: 178,1631 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00276110
CAS nummer: 1735-88-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR