Diphenyl(trifluoromethyl)sulphonium trifluoromethanesulphonate 98%
Leverantör: Apollo Scientific
|
Varning
|
PC905034-1G
PC905034-250MG
PC905034-5G
APOSPC905034-1GEA
3075
SEK
APOSPC905034-1G
APOSPC905034-250MG
APOSPC905034-5G
Diphenyl(trifluoromethyl)sulphonium trifluoromethanesulphonate 98%
Diphenyl(trifluoromethyl)sulphonium trifluoromethanesulphonate
Formel:
C₁₄H₁₀F₆O₃S₂ MW: 404,35 g/mol Storage Temperature: Omgivande |
MDL Number:
MFCD12912173 CAS nummer: 147531-11-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...