Methyl-2-amino-3,6-difluorobenzoate 95%
Leverantör: Apollo Scientific
Synonyms: Methyl 2-amino-3,6-difluorobenzoate
PC903670-1G PC903670-5G
APOSPC903670-1GEA 2500 SEK
APOSPC903670-1G APOSPC903670-5G
Methyl-2-amino-3,6-difluorobenzoate 95%
Methyl-2-amino-3,6-difluorobenzoate
Formel: C₈H₇F₂NO₂
MW: 187,1400 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD12159870
CAS nummer: 1184204-30-5