1-Bromo-5-chloro-3-fluoro-2-iodobenzene 98%
Leverantör: Apollo Scientific
Varning
PC902929-100G PC902929-25G
APOSPC902929-100GEA 4275 SEK
APOSPC902929-100G APOSPC902929-25G
1-Bromo-5-chloro-3-fluoro-2-iodobenzene 98%
1-Bromo-5-chloro-3-fluoro-2-iodobenzene
Formel: C₆H₂BrClFI
Storage Temperature: Omgivande
MDL Number: MFCD00142591
CAS nummer: 201849-16-3