3-Fluoro-4-methoxythiophenol 95%

Leverantör: Apollo Scientific

PC902760-1G PC902760-250MG
APOSPC902760-1GEA 3600 SEK
APOSPC902760-1G APOSPC902760-250MG
3-Fluoro-4-methoxythiophenol 95%
3-Fluoro-4-methoxythiophenol
Formel: C₇H₇FOS
Storage Temperature: Omgivande
MDL Number: MFCD06201751
CAS nummer: 89818-27-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR