6-Fluoro-2-methylnicotinic acid 95%

Leverantör: Apollo Scientific

Synonyms: 6-Fluoro-2-methyl-3-pyridinecarboxylic acid

PC900652-100MG PC900652-1G PC900652-250MG
APOSPC900652-100MGEA 1725 SEK
APOSPC900652-100MG APOSPC900652-1G APOSPC900652-250MG
6-Fluoro-2-methylnicotinic acid 95%
6-Fluoro-2-methylnicotinic acid
Formel: C₇H₆FNO₂
MW: 155,1300 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD05662400
CAS nummer: 884494-97-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR