2-Hydrazino-6-(trifluoromethyl)pyridine
Leverantör: Apollo Scientific
Varning
PC8991-1G PC8991-250MG PC8991-5G
APOSPC8991-1GEA 1925 SEK
APOSPC8991-1G APOSPC8991-250MG APOSPC8991-5G
2-Hydrazino-6-(trifluoromethyl)pyridine
2-Hydrazino-6-(trifluoromethyl)pyridine
Formel: C₆H₆F₃N₃
Storage Temperature: Kylskåp
MDL Number: MFCD00067866
CAS nummer: 94239-06-2