1-[6-(Trifluoromethyl)pyridin-2-yl]piperidine-4-carboxylic acid
Leverantör: Apollo Scientific
PC8978-1G PC8978-10MG PC8978-50MG PC8978-100MG PC8978-250MG PC8978-500MG
APOSPC8978-1GEA 3250 SEK
APOSPC8978-1G APOSPC8978-10MG APOSPC8978-50MG APOSPC8978-100MG APOSPC8978-250MG APOSPC8978-500MG
1-[6-(Trifluoromethyl)pyridin-2-yl]piperidine-4-carboxylic acid
Formel: C₁₂H₁₃F₃N₂O₂
Storage Temperature: Omgivande
MDL Number: MFCD08235221
CAS nummer: 582325-39-1