2-Chloro-3-fluoroisonicotinic acid

Leverantör: Apollo Scientific
Varning

Synonyms: 2-Chloro-3-fluoropyridine-4-carboxylic acid

PC8963-1G
APOSPC8963-1GEA 260 SEK
APOSPC8963-1G
2-Chloro-3-fluoroisonicotinic acid
2-Chloro-3-fluoroisonicotinic acid
Formel: C₆H₃ClFNO₂
MW: 175.55 g/mol
Smältpunkt: 199 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD04038325
CAS nummer: 628691-93-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR