3-Chloro-5-(trifluoromethyl)picolinonitrile
Leverantör: Apollo Scientific
Varning
Synonyms: 3-Chloro-5-(trifluoromethyl)pyridine-2-carbonitrile
PC8824-1G PC8824-5G PC8824-10G
APOSPC8824-1GEA 175 SEK
APOSPC8824-1G APOSPC8824-5G APOSPC8824-10G
3-Chloro-5-(trifluoromethyl)picolinonitrile
3-Chloro-5-(trifluoromethyl)picolinonitrile
Formel: C₇H₂ClF₃N₂
MW: 206,5524 g/mol
Kokpunkt: 90 °C/11mmHg
Storage Temperature: Kylskåp
MDL Number: MFCD00664066
CAS nummer: 80194-70-3