2-Methyl-4-(trifluoromethoxy)anisole 97%

Leverantör: Apollo Scientific

Synonyms: 2-Methoxy-5-(trifluoromethoxy)toluene

PC8762-1G PC8762-5G
APOSPC8762-1GEA 2125 SEK
APOSPC8762-1G APOSPC8762-5G
2-Methyl-4-(trifluoromethoxy)anisole 97%
2-Methyl-4-(trifluoromethoxy)anisole
Formel: C₉H₉F₃O₂
Storage Temperature: Omgivande
MDL Number: MFCD04972926
CAS nummer: 887266-88-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR