4-[3-(Trifluoromethyl)phenyl]piperidine hydrochloride 97%
Leverantör: Apollo Scientific
Varning
Synonyms: 4-(3-(Trifluoromethyl)phenyl)piperidine hydrochloride
PC8686-1G PC8686-5G
APOSPC8686-1GEA 600 SEK
APOSPC8686-1G APOSPC8686-5G
4-[3-(Trifluoromethyl)phenyl]piperidine hydrochloride 97%
4-[3-(Trifluoromethyl)phenyl]piperidine hydrochloride
Formel: C₁₂H₁₅ClF₃N
MW: 265.71 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD02178913
CAS nummer: 6652-16-0