2-Chloro-6-fluorobenzaldehyde oxime, Technical Grade
Leverantör: Apollo Scientific
Varning
Synonyms: 2-Chloro-6-fluorobenzaldoxime
PC8674-1G PC8674-5G
APOSPC8674-1GEA 415 SEK
APOSPC8674-1G APOSPC8674-5G
2-Chloro-6-fluorobenzaldehyde oxime, Technical Grade
2-Chloro-6-fluorobenzaldehyde oxime
Formel: C₇H₅ClFNO
MW: 173,57 g/mol
Smältpunkt: 133…134 °C
Storage Temperature: Omgivande
MDL Number: MFCD00837704
CAS nummer: 443-33-4