7-(Trifluoromethyl)-1-benzothiophene-2-carboxylic acid
Leverantör: Apollo Scientific
Varning
Synonyms: 7-(Trifluoromethyl)benzo[b]thiophene-2-carboxylic acid
PC8617-1G PC8617-5G PC8617-10G
APOSPC8617-1GEA 1125 SEK
APOSPC8617-1G APOSPC8617-5G APOSPC8617-10G
7-(Trifluoromethyl)-1-benzothiophene-2-carboxylic acid
7-(Trifluoromethyl)-1-benzothiophene-2-carboxylic acid
Formel: C₁₀H₅F₃O₂S
MW: 246,21 g/mol
Smältpunkt: 191…195 °C
Storage Temperature: Omgivande
MDL Number: MFCD09027110
CAS nummer: 550998-66-8