Pentafluorophenyl-(4-methyl-6,7-dihydro-4H-thieno[3,2-c]pyran-4-yl)acetate 97%
Leverantör: Apollo Scientific
PC8548-1G PC8548-250MG
APOSPC8548-1GEA 2350 SEK
APOSPC8548-1G APOSPC8548-250MG
Pentafluorophenyl-(4-methyl-6,7-dihydro-4H-thieno[3,2-c]pyran-4-yl)acetate 97%
Pentafluorophenyl-(4-methyl-6,7-dihydro-4H-thieno[3,2-c]pyran-4-yl)acetate
Formel: C₁₆H₁₁F₅O₃S
MW: 378,32 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD09817505
CAS nummer: 916766-89-7