2-Bromo-2'-fluoro-6'-(trifluoromethyl)acetophenone 97%
Leverantör: Apollo Scientific
Fara
Synonyms: 2-Fluoro-6-(trifluoromethyl)phenacyl bromide
PC8404-1G PC8404-5G
APOSPC8404-1GEA 1125 SEK
APOSPC8404-1G APOSPC8404-5G
2-Bromo-2'-fluoro-6'-(trifluoromethyl)acetophenone 97%
2-Bromo-2'-fluoro-6'-(trifluoromethyl)acetophenone
Formel: C₉H₅BrF₄O
Storage Temperature: Kylskåp
MDL Number: MFCD03094503
CAS nummer: 223785-85-1
UN: 1760
ADR: 8 III