N-Boc-2-trifluoromethyl-L-phenylalanine
Leverantör: Apollo Scientific
Varning
Synonyms: (S)-2-((tert-Butoxycarbonyl)amino)-3-(2-(trifluoromethyl)phenyl)propanoic acid, 2-(Trifluoromethyl)-L-phenylalanine, N-BOC protected
PC8354-1G PC8354-25G PC8354-5G
APOSPC8354-1GEA 1950 SEK
APOSPC8354-1G APOSPC8354-25G APOSPC8354-5G
N-Boc-2-trifluoromethyl-L-phenylalanine
N-Boc-2-trifluoromethyl-L-phenylalanine
Formel: C₁₅H₁₈F₃NO₄
MW: 333.307 g/mol
Storage Temperature: Frys
MDL Number: MFCD01862946
CAS nummer: 167993-21-7