Bromotrispyrrolidinophosphonium hexafluorophosphate
Leverantör: Apollo Scientific
Fara
Synonyms: Bromotri(1-pyrrolidinyl)phosphonium hexafluorophosphate, PyBrOP , Bromotripyrrolidinylphosphonium hexafluorophosphate, Bromotri(pyrrolidin-1-yl)phosphonium hexafluorophosphate(V)
PC8353-100G PC8353-250G PC8353-25G
APOSPC8353-100GEA 2350 SEK
APOSPC8353-100G APOSPC8353-250G APOSPC8353-25G
Bromotrispyrrolidinophosphonium hexafluorophosphate
Bromotrispyrrolidinophosphonium hexafluorophosphate
Formel: C₁₂H₂₄BrF₆N₃P₂
MW: 466,2 g/mol
Smältpunkt: 139 °C
Storage Temperature: Frys
MDL Number: MFCD00077412
CAS nummer: 132705-51-2