1-Bromo-2,3,6-trifluorobenzen 98%

Leverantör: Apollo Scientific
Varning

PC8320-1G PC8320-5G PC8320-25G
APOSPC8320-1GEA 175 SEK
APOSPC8320-1G APOSPC8320-5G APOSPC8320-25G
1-Bromo-2,3,6-trifluorobenzen 98%
1-Bromo-2,3,6-trifluorobenzen
Formel: C₆H₂BrF₃
MW: 210,98 g/mol
Flampunkt: 62 °C
Storage Temperature: Omgivande
MDL Number: MFCD00236214
CAS nummer: 176793-04-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR