Methyl-4-chloro-2-fluorobenzoate
Leverantör: Apollo Scientific
Varning
Synonyms: Methyl 4-chloro-2-fluorobenzoate, 4-Chloro-2-fluorobenzoic acid methyl ester
PC8037-25G PC8037-5G PC8037-1G
APOSPC8037-25GEA 3025 SEK
APOSPC8037-25G APOSPC8037-5G APOSPC8037-1G
Methyl-4-chloro-2-fluorobenzoate
Methyl-4-chloro-2-fluorobenzoate
Formel: C₈H₆ClFO₂
MW: 188,59 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD04038392
CAS nummer: 148893-72-5