2,4,6-Tris(pentafluoroethyl)-1,3,5-triazine
Leverantör: Apollo Scientific
Varning
Synonyms: 2,4,6-Tris(perfluoroethyl)-1,3,5-triazine
PC7900-1G PC7900-250MG PC7900-5G
APOSPC7900-1GEA 1525 SEK
APOSPC7900-1G APOSPC7900-250MG APOSPC7900-5G
2,4,6-Tris(pentafluoroethyl)-1,3,5-triazine
2,4,6-Tris(pentafluoroethyl)-1,3,5-triazine
Mass spectrometry standard.
Formel: C₉F₁₅N₃
MW: 435.10 g/mol
Kokpunkt: 121…122 °C
Densitet: 1.6506 g/cm³ (25 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00042437
CAS nummer: 858-46-8
EINECS: 212-724-6
Order Now
SPECIFIKATIONER
Environmentally Preferable