4,6-Difluoroindole 96%
Leverantör: Apollo Scientific
Varning
PC7811-1G PC7811-5G
APOSPC7811-1GEA 685 SEK
APOSPC7811-1G APOSPC7811-5G
4,6-Difluoroindole 96%
4,6-Difluoroindole
Formel: C₈H₅F₂N
MW: 153,13 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD01075213
CAS nummer: 199526-97-1