4-Bromo-2,6-difluorophenylboronic acid
Leverantör: Apollo Scientific
|
Varning
|
Synonyms:
4-Bromo-2,6-difluorobenzeneboronic acid
PC7580-1G
PC7580-5G
APOSPC7580-1GEA
585
SEK
APOSPC7580-1G
APOSPC7580-5G
4-Bromo-2,6-difluorophenylboronic acid
4-Bromo-2,6-difluorophenylboronic acid
Formel:
C₆H₄BBrF₂O₂ MW: 236,807 g/mol Smältpunkt: 155…160 °C Storage Temperature: Kylskåp |
MDL Number:
MFCD03095369 CAS nummer: 352535-81-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...