4-Bromo-2-nitro-5-(trifluoromethyl)aniline
Leverantör: Apollo Scientific
|
Varning
|
Synonyms:
5-Amino-2-bromo-4-nitrobenzotrifluoride
PC7517-1G
PC7517-250MG
APOSPC7517-1GEA
1325
SEK
APOSPC7517-1G
APOSPC7517-250MG
4-Bromo-2-nitro-5-(trifluoromethyl)aniline
4-Bromo-2-nitro-5-(trifluoromethyl)aniline
Formel:
C₇H₄BrF₃N₂O₂ MW: 285,0181 g/mol Smältpunkt: 70…74 °C Flampunkt: >110 °C Storage Temperature: Kylskåp |
MDL Number:
MFCD09263997 CAS nummer: 683241-86-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...