3-Chloro-2-iodo-5-trifluoromethylpyridine

Leverantör: Apollo Scientific
Varning

PC7479-1G PC7479-5G
APOSPC7479-1GEA 855 SEK
APOSPC7479-1G APOSPC7479-5G
3-Chloro-2-iodo-5-trifluoromethylpyridine
3-Chloro-2-iodo-5-trifluoromethylpyridine
Formel: C₆H₂ClF₃IN
MW: 307,44 g/mol
Smältpunkt: 39…41 °C
Storage Temperature: Omgivande
MDL Number: MFCD09264543
CAS nummer: 134161-12-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR