Trifluoromethanesulphonyl chloride 97%
Leverantör: Apollo Scientific
Fara
PC7430-100G PC7430-25G PC7430-250G
APOSPC7430-100GEA 6425 SEK
APOSPC7430-100G APOSPC7430-25G APOSPC7430-250G
Trifluoromethanesulphonyl chloride 97%
Trifluoromethanesulphonyl chloride
Formel: CClF₃O₂S
MW: 168,52 g/mol
Kokpunkt: 29…32 °C
Densitet: 1,583 g/cm³ (25 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00007451
CAS nummer: 421-83-0
EINECS: 207-009-0
UN: 3265
ADR: 8,II