2,2,2-Trifluoroethyl trifluoroacetate 99%
Leverantör: Apollo Scientific
Fara
Synonyms: Trifluoroacetic acid (2,2,2-trifluoroethyl) ester
PC7378-100G PC7378-500G
APOSPC7378-100GEA 4925 SEK
APOSPC7378-100G APOSPC7378-500G
2,2,2-Trifluoroethyl trifluoroacetate 99%
2,2,2-Trifluoroethyl trifluoroacetate
Formel: C₄H₂F₆O₂
Kokpunkt: 55 °C
Densitet: 1,464 g/cm³ (25 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00000418
CAS nummer: 407-38-5
EINECS: 206-985-5
UN: 2924
ADR: 3+8,II