Pentafluorophenyl trifluoromethanesulphonate
Leverantör: Apollo Scientific
Fara
Synonyms: Trifluoromethanesulphonic acid Pentafluorophenyl Ester, Perfluorophenyl trifluoromethanesulphonate, Pentafluorophenyl Triflate
PC7353-1G PC7353-25G PC7353-5G
APOSPC7353-1GEA 945 SEK
APOSPC7353-1G APOSPC7353-25G APOSPC7353-5G
Pentafluorophenyl trifluoromethanesulphonate
Pentafluorophenyl trifluoromethanesulphonate
Formel: C₇F₈O₃S
MW: 316,12 g/mol
Kokpunkt: 81 °C/38mmHg
Flampunkt: 89 °C
Storage Temperature: Omgivande
MDL Number: MFCD07784319
CAS nummer: 60129-85-3
UN: 3265
ADR: 8,III