Trifluorättiksyra 99,9%, Biochemical Grade
Leverantör: Apollo Scientific
Fara
Synonyms: TFA
PC7161-1KG PC7161-250G PC7161-5KG
APOSPC7161-1KGEA 925 SEK
APOSPC7161-1KG APOSPC7161-250G APOSPC7161-5KG
Trifluorättiksyra 99,9%, Biochemical Grade
Trifluorättiksyra
Suitable for protein sequence analysis.
Formel: C₂HF₃O₂
Kokpunkt: 72,4 °C
Densitet: 1,489 g/cm³ (25 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00004169
CAS nummer: 76-05-1
EINECS: 200-929-3
UN: 2699
ADR: 8,I